missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134887
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P09905G 5 g
P099025G 25 g
2 options

Catalog No. P09905G

Supplier: TCI America P09905G

Only null left

Chemical Identifiers

CAS 15430-52-1
Molecular Formula C3H6ClN
Molecular Weight (g/mol) 91.54
MDL Number MFCD00012907
InChI Key IKXNIQJDNKPPCH-UHFFFAOYSA-N
Synonym propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
PubChem CID 11205720
IUPAC Name hydrogen prop-2-yn-1-amine chloride
SMILES [H+].[Cl-].NCC#C

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 91.54
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Propargylamine Hydrochloride