Learn More
Propargylamine hydrochloride, 95%
CAS: 15430-52-1 | C3H6ClN | 91.54 g/mol
$76.63 - $204.78
Chemical Identifiers
| CAS | 15430-52-1 |
|---|---|
| Molecular Formula | C3H6ClN |
| Molecular Weight (g/mol) | 91.54 |
| MDL Number | MFCD00012907 |
| InChI Key | IKXNIQJDNKPPCH-UHFFFAOYSA-N |
| Synonym | propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh |
| PubChem CID | 11205720 |
| IUPAC Name | prop-2-yn-1-amine;hydrochloride |
| SMILES | [H+].[Cl-].NCC#C |
Description
Propargylamine hydrochloride is a useful intermediates in the preparation of drugs. It is used in pharmaceutical, industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15430-52-1 | |
| 91.54 | |
| IKXNIQJDNKPPCH-UHFFFAOYSA-N | |
| 11205720 | |
| [H+].[Cl-].NCC#C |
| C3H6ClN | |
| MFCD00012907 | |
| propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh | |
| prop-2-yn-1-amine;hydrochloride |
Specifications
| 15430-52-1 | |
| C3H6ClN | |
| 1 g | |
| Hygroscopic | |
| Soluble in water (partly),methanol,and ethanol. | |
| [H+].[Cl-].NCC#C | |
| 91.54 | |
| 91.54 | |
| Powder |
| 179°C to 182°C | |
| MFCD00012907 | |
| UN2811 | |
| propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh | |
| IKXNIQJDNKPPCH-UHFFFAOYSA-N | |
| prop-2-yn-1-amine;hydrochloride | |
| 11205720 | |
| 95% | |
| Propargylamine hydrochloride |
Safety and Handling
GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 239-440-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only