missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propargylamine hydrochloride, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 175°C to 182°C |
| Assay Percent Range | 94% min. (Argentometry) |
| Packaging | Glass bottle |
| Linear Formula | HC∽CCH2NH2·HCl |
| Quantity | 10 g |
| Beilstein | 04, 228 |
| Formula Weight | 91.54 |
| Percent Purity | 95% |
| Chemical Name or Material | Propargylamine hydrochloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Harmful if swallowed. Harmful in contact with skin. May cause respiratory irritation. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor |
| EINECSNumber | 239-440-5 |
RUO – Research Use Only