missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine hydrochloride, 95%

Catalog No. AA4377503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4377503 1 g
AA4377506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4377503 Supplier Thermo Scientific Chemicals Supplier No. 04377503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15430-52-1 | C3H6ClN | 91.54 g/mol

Propargylamine hydrochloride is a useful intermediates in the preparation of drugs. It is used in pharmaceutical, industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15430-52-1
Molecular Formula C3H6ClN
Molecular Weight (g/mol) 91.54
MDL Number MFCD00012907
InChI Key IKXNIQJDNKPPCH-UHFFFAOYSA-N
Synonym propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
PubChem CID 11205720
IUPAC Name prop-2-yn-1-amine;hydrochloride
SMILES [H+].[Cl-].NCC#C

Specifications

Melting Point 179°C to 182°C
Quantity 1 g
UN Number UN2811
Sensitivity Hygroscopic
Solubility Information Soluble in water (partly),methanol,and ethanol.
Formula Weight 91.54
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Propargylamine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 239-440-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only