missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine, 99%

Catalog No. p-187716
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC131460050 5 g
AC131460250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131460050 Supplier Thermo Scientific Chemicals Supplier No. 131460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2450-71-7 | C3H5N | 55.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00008198
InChI Key JKANAVGODYYCQF-UHFFFAOYSA-N
Synonym propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine
PubChem CID 239041
IUPAC Name prop-2-yn-1-amine
SMILES C#CCN

Specifications

Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 84.0°C
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4500
Linear Formula HCidentical toCCH2NH2
Quantity 5 g
Beilstein 04,228
Specific Gravity 0.86
Solubility Information Solubility in water: miscible in water.
Formula Weight 55.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propargylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gentl
EINECSNumber 219-513-8
RTECSNumber UK5250000
TSCA TSCA

RUO – Research Use Only