missing translation for 'onlineSavingsMsg'
Learn More

Propargyl p-Toluenesulfonate 96.0+%, TCI America™

Catalog No. T14555G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14555G Supplier TCI America Supplier No. T14555G

Chemical Identifiers

CAS 6165-76-0
Molecular Formula C10H10O3S
Molecular Weight (g/mol) 210.247
MDL Number MFCD00078365
InChI Key LMBVCSFXFFROTA-UHFFFAOYSA-N
Synonym propargyl p-toluenesulfonate, propargyl tosylate, prop-2-yn-1-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid, 2-propynyl ester, 2-propyn-1-ol, 4-methylbenzenesulfonate, ccris 9146, p-toluenesulfonic acid propargyl ester, propargylp-toluenesulfonate, propargyl p-toluene sulfonate, acmc-209mv7
PubChem CID 22547
IUPAC Name prop-2-ynyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC#C

Specifications

Quantity 5 g
Formula Weight 210.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargyl p-Toluenesulfonate
RTECSNumber XT7300000
TSCA No