missing translation for 'onlineSavingsMsg'
Learn More

Propargyl p-toluenesulfonate, 97%

Catalog No. AAL1207509
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1207509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1207509 Supplier Thermo Scientific Chemicals Supplier No. L1207509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6165-76-0 | C10H10O3S | 210.247 g/mol

Propargyl p-toluenesulfonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl p-toluenesulfonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Chloroform, Dichloromethane, DMSO, Ethyl Aceta

Chemical Identifiers

CAS 6165-76-0
Molecular Formula C10H10O3S
Molecular Weight (g/mol) 210.247
MDL Number MFCD00078365
InChI Key LMBVCSFXFFROTA-UHFFFAOYSA-N
Synonym propargyl p-toluenesulfonate, propargyl tosylate, prop-2-yn-1-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid, 2-propynyl ester, 2-propyn-1-ol, 4-methylbenzenesulfonate, ccris 9146, p-toluenesulfonic acid propargyl ester, propargylp-toluenesulfonate, propargyl p-toluene sulfonate, acmc-209mv7
PubChem CID 22547
IUPAC Name prop-2-ynyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC#C

Specifications

Density 1.22
Boiling Point 117°C to 121°C (0.3 mmHg)
Refractive Index 1.527
Quantity 10 g
Beilstein 1212957
Solubility Information Soluble in Chloroform,Dichloromethane,DMSO,Ethyl Acetate. Not miscible in water.
Formula Weight 210.25
Percent Purity 97%
Chemical Name or Material Propargyl p-toluenesulfonate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber XT7600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only