Learn More
Propargyl p-toluenesulfonate, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 6165-76-0 |
|---|---|
| Molecular Formula | C10H10O3S |
| Molecular Weight (g/mol) | 210.25 |
| MDL Number | MFCD01462194 |
| InChI Key | LMBVCSFXFFROTA-UHFFFAOYSA-N |
| Synonym | propargyl p-toluenesulfonate, propargyl tosylate, prop-2-yn-1-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid, 2-propynyl ester, 2-propyn-1-ol, 4-methylbenzenesulfonate, ccris 9146, p-toluenesulfonic acid propargyl ester, propargylp-toluenesulfonate, propargyl p-toluene sulfonate, acmc-209mv7 |
| PubChem CID | 22547 |
| IUPAC Name | prop-2-ynyl 4-methylbenzenesulfonate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCC#C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC379680050
|
Thermo Scientific Chemicals
379680050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC379680250
|
Thermo Scientific Chemicals
379680250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC379681000
|
Thermo Scientific Chemicals
379681000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 6165-76-0 | |
| 210.25 | |
| LMBVCSFXFFROTA-UHFFFAOYSA-N | |
| 22547 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCC#C |
| C10H10O3S | |
| MFCD01462194 | |
| propargyl p-toluenesulfonate, propargyl tosylate, prop-2-yn-1-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid, 2-propynyl ester, 2-propyn-1-ol, 4-methylbenzenesulfonate, ccris 9146, p-toluenesulfonic acid propargyl ester, propargylp-toluenesulfonate, propargyl p-toluene sulfonate, acmc-209mv7 | |
| prop-2-ynyl 4-methylbenzenesulfonate |
Specifications
| 6165-76-0 | |
| 96% min. (GC) | |
| C10H10O3S | |
| 1.5270 to 1.5290 (20°C, 589nm) | |
| 5g | |
| propargyl p-toluenesulfonate, propargyl tosylate, prop-2-yn-1-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid, 2-propynyl ester, 2-propyn-1-ol, 4-methylbenzenesulfonate, ccris 9146, p-toluenesulfonic acid propargyl ester, propargylp-toluenesulfonate, propargyl p-toluene sulfonate, acmc-209mv7 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCC#C | |
| 210.25 | |
| 210.25 | |
| Propargyl p-toluenesulfonate |
| Authentic | |
| Glass bottle | |
| CH3C6H4SO3CH2C≡CH | |
| MFCD01462194 | |
| 11, IV, 263 | |
| LMBVCSFXFFROTA-UHFFFAOYSA-N | |
| prop-2-ynyl 4-methylbenzenesulfonate | |
| 22547 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only