missing translation for 'onlineSavingsMsg'
Learn More

Propargyl chloride, 98%

Catalog No. p-4813628
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC451840050 5 g Glass bottle
AC451840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451840050 Supplier Thermo Scientific Chemicals Supplier No. 451840050

CAS: 624-65-7 | C3H3Cl | 74.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-65-7
Molecular Formula C3H3Cl
Molecular Weight (g/mol) 74.51
MDL Number MFCD00000980
InChI Key LJZPPWWHKPGCHS-UHFFFAOYSA-N
Synonym propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
PubChem CID 12221
IUPAC Name 3-chloroprop-1-yne
SMILES ClCC#C

Specifications

Density 1.0300g/mL
Boiling Point 58.0°C to 61.0°C
Flash Point 19°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2Cl
Quantity 5 g
Merck Index 15,7924
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble.
Formula Weight 74.51
Percent Purity 98%
Chemical Name or Material Propargyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
EINECSNumber 210-856-9

RUO – Research Use Only