missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Bromide (stabilized with MgO) 97.0+%, TCI America™

Catalog No. P0484500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0484500G 500 g
P048425G 25 g
P0484100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0484500G Supplier TCI America Supplier No. P0484500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-96-7
Molecular Formula C3H3Br
Molecular Weight (g/mol) 118.961
MDL Number MFCD00000241
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
PubChem CID 7842
IUPAC Name 3-bromoprop-1-yne
SMILES C#CCBr

Specifications

Color Yellow
Boiling Point 90°C
Quantity 500 g
UN Number 2345
Formula Weight 118.96
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargyl Bromide (stabilized with MgO)
EINECSNumber (2)-0108
RTECSNumber UK4375000
TSCA Yes
Recommended Storage Refrigerator