missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™

CAS: 106-96-7 | C3H3Br | 118.96 g/mol

$260.03 - $260.03

Specifications

Boiling Point 88.0°C to 90.0°C
Chemical Name or Material Propargyl bromide
Name Note Stabilized
CAS 106-96-7,108-88-3
Color White to Yellow
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC429121000
View Documents
Thermo Scientific Chemicals
429121000
100 mL AcroSeal™ Glass Bottle
Each for $260.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

88.0°C to 90.0°C
Stabilized
White to Yellow
C3H3Br
MFCD00000241
propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
YORCIIVHUBAYBQ-UHFFFAOYSA-N
3-bromoprop-1-yne
7842
73 to 87% (propargyl bromide) (GC), 13 to 27% (toluene) (GC)
1.4900 to 1.4960
1.3800g/mL
Propargyl bromide
106-96-7,108-88-3
Crystalline Powder
HC≡CCH2Br
4°C
Solubility in water: immiscible
C#CCBr
118.96
118.96
AcroSeal™ Glass Bottle
1.38
Videos
SDS
Documents

Documents

RUO – Research Use Only