missing translation for 'onlineSavingsMsg'
Learn More

Propargyl acrylate, 96%, stab. with ca 200ppm BHT

Catalog No. AAL0938309
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0938309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0938309 Supplier Thermo Scientific Chemicals Supplier No. L0938309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10477-47-1 | C6H6O2 | 110.11 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 10477-47-1
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00078451
InChI Key WPBNLDNIZUGLJL-UHFFFAOYSA-N
Synonym propargyl acrylate, 2-propenoic acid, 2-propynyl ester, acrylic acid propargyl ester, 2-propenoic acid, 2-propyn-1-yl ester, 2-propynyl acrylate, prop-2-yn-1-yl acrylate, acmc-1c257, prop-2-yn-1-yl prop-2-enoate, propargyl acrylate,
PubChem CID 82655
SMILES C=CC(=O)OCC#C

Specifications

Density 0.995
Boiling Point 138°C
Flash Point 43°C (109°F)
Odor Pungent
Refractive Index 1.446
Linear Formula H2C=CHCO2CH2C≡CH
Quantity 10 g
UN Number UN3272
Beilstein 1902961
Solubility Information Not miscible or difficult to mix with water.
IUPAC Name prop-2-ynyl prop-2-enoate
Formula Weight 110.11
Percent Purity 96%
Chemical Name or Material Propargyl acrylate, stabilized with ≈200ppm BHT
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 233-975-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only