missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propargyl acetate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.989 |
| Boiling Point | 124°C |
| Flash Point | 31°C (88°F) |
| Odor | Characteristic |
| Refractive Index | 1.4185 |
| Linear Formula | CH3CO2CH2C≡CH |
| Quantity | 5 g |
| UN Number | UN1992 |
| Beilstein | 1742046 |
| Formula Weight | 98.1 |
| Show More |
Safety and Handling
| Hazard Category | H226-H301-H312+H332 |
| Hazard Statement | GHS H Statement H301-H226 Toxic if swallowed. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
| RTECSNumber | UK5076000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only