missing translation for 'onlineSavingsMsg'
Learn More

Propargyl acetate, 97%

Catalog No. AAL1003114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1003114 25 g
AAL1003106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1003114 Supplier Thermo Scientific Chemicals Supplier No. L1003114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-09-8 | C5H6O2 | 98.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-09-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00041601
InChI Key RIZZXCJMFIGMON-UHFFFAOYSA-N
Synonym propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
PubChem CID 69388
IUPAC Name prop-2-ynyl acetate
SMILES CC(=O)OCC#C

Specifications

Density 0.989
Boiling Point 124°C
Flash Point 31°C (88°F)
Odor Characteristic
Refractive Index 1.4185
Linear Formula CH3CO2CH2C≡CH
Quantity 25 g
UN Number UN1992
Beilstein 1742046
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material Propargyl acetate
Show More Show Less
Hazard Category H226-H301-H312+H332
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
RTECSNumber UK5076000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only