missing translation for 'onlineSavingsMsg'
Learn More

Promega ΦX174, RF DNA

Numéro de catalogue. PRD1531
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PRD1531 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PRD1531 Fournisseur Promega Code fournisseur D1531
Il en reste null

phiX174 replicative form (RF) is a double-stranded circular DNA molecule of 5,386 bases. Used in assays of restriction enzymes for the presence of nickase activity.

  • Double-Stranded Circular DNA Molecule Used to Detect Restriction Enzyme Assay Nickase Activity
  • Supplied at a concentration of 1mg/ml

Spécifications

Concentration 1 μg/μL
Form Liquid
For Use With (Application) For used as a substrate in restriction Enzyme activity assays
Quantity 50 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Standard