Learn More
Promega Wheat Germ Extract
Wheat Germ Extract contains the cellular components necessary for protein synthesis (tRNA, ribosomes, initiation, elongation and termination factors) and is used for in vitro translation..
Supplier: Promega L4380
Includes:
Wheat germ extract, 5 x 200μL; BMV RNA, 5μg; Potassium acetate, 0.5mL; Amino acid mixtures, 3 x 175μL
Description
- Components for Protein Synthesis
- Contains tRNA, ribosomes, initiation, elongation and termination factors
- Extract is cleared of endogenous amino acids, mRNA and plant pigments, minimizing background translation
Specifications
| <-65°C | |
| Expression of eukaryotic messages that do not express well in reticulocyte lysates, expression of plant mRNAs, expression of mammalian transcription factors | |
| 5 x 200μL Wheat Germ extract, 5μg BMV RNA, 0.5mL Potassium Acetate, 3 x 175μL Amino Acid mixtures | |
| 5 x 200 μL |