missing translation for 'onlineSavingsMsg'
Learn More

Promega Ultra Pure Deoxynucleotide Triphosphates (dNTPs): PCR Nucleotide Mix

PCR Nucleotide Mix is a premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP, each at a concentration of 10mM or 25mM in water.

$135.71 - $534.95

Specifications

pH 7.5
Content And Storage -20°C
Concentration 10 mM
Purity >98%
For Use With (Application) cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRC1141
Encompass
View Documents Promotion Details
Promega
C1141
200 μL
Each for $135.71
Only null left
 
PRC1145
Encompass
View Documents Promotion Details
Promega
C1145
1000 μL
Each for $534.95
Only null left
 
Description

Description

  • cGMP-Grade Solution of dNTPs Pretested in PCR
  • cGMP-manufactured for lot-to-lot product consistency
  • Equimolar amounts of each dNTP in water
  • Two concentrations available: 10mM and 25mM
Specifications

Specifications

7.5
10 mM
cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols
-20°C
>98%
Videos
SDS
Documents

Documents