missing translation for 'onlineSavingsMsg'
Learn More

Promega Disposable Polypropylene Test Tubes

Catalog No. PRE4221
Encompass
Change view
Click to view available options
Type:
Test Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
PRE4221 Test Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE4221 Supplier Promega Supplier No. E4221

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This accessory is available for the now-discontinued 20/20 luminometer.

  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued

Specifications

Type Test Tube
For Use With (Equipment) Turner BioSystems Luminometer Model TD-20/20
Includes 1000/Pk
Storage Requirements 15 to 30°C