missing translation for 'onlineSavingsMsg'
Learn More

Promega Interface Tubing and Valves for Auto Injector System

Catalog No. PRE2211
Encompass
Change view
Click to view available options
For Use With (Equipment):
Turner BioSystems Luminometer Model TD-20/20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
PRE2211 Turner BioSystems Luminometer Model TD-20/20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE2211 Supplier Promega Supplier No. E2211

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This accessory are available for the now-discontinued 20/20 luminometer.

  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued

Specifications

Type Interface Tubing and Valves for Auto Injector System
For Use With (Equipment) Turner BioSystems Luminometer Model TD-20/20
Includes 1/ea
Storage Requirements 15 to 30°C