missing translation for 'onlineSavingsMsg'
Learn More

Promega 5X Transcription Optimized Buffer

Catalog No. PRP1181
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRP1181 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRP1181 Supplier Promega Supplier No. P1181

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Riboprobe System Buffers are qualified for use in applications where maintaining the integrity of RNA is critical. Product is quality tested to ensure the absence of detectable RNase activity.

  • Package of Transcription-Optimized Buffer, rNTPs and Nuclease-Free Water
  • Used with Riboprobe(R) Systems
  • Qualified for use for in vitro transcription reactions with SP6, T7 or T3 RNA polymerase
  • Reagents tested with other Riboprobe(R) System components. rNTPs tested for functionality with in vitro transcription reactions.

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Used as a components of the single and combination Riboprobe™ systems
Includes RQ1 RNase-free Dnase, rATP, rCTP, rGTP and rUTP, 10X Reaction Buffer and 10X Stop Buffer
Quantity 200 μL