missing translation for 'onlineSavingsMsg'
Learn More

Promega Transcend™ Nonradioactive Translation Detection Systems

Catalog No. PRL5070
Encompass
Change view
Click to view available options
Quantity:
30 x 50 μL Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL5070 30 x 50 μL Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRL5070 Supplier Promega Supplier No. L5070

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-Radioactive Translation Detection Systems for colorimetric or chemiluminescent detection of proteins synthesized in vitro. Biotinylated proteins are detected using Streptavidin-Alkaline Phosphatase or Streptavidin-HRP.

  • Non-Radioactive Detection of In Vitro Translated Proteins
  • Biotin tag allows detection of 0.5-5ng of protein 3-4 hours after gel electrophoresis
  • No radioisotope handling, storage or disposal issues
  • Option of colorimetric or chemiluminescent detection

Specifications

Content And Storage COMPONENTS AT DIFFERENT TEMPS
For Use With (Application) Non-radioactive detection of Proteins synthesized in-vitro
Includes 30μL Transcend™ Trna, 36μL Streptavidin-AP, 35mL Western Blue™ stabilized substrate for Alkaline Phosphatase
Product Type Transcend Colorimetric Nonradioactive Translation Detection System
Quantity 30 x 50 μL Reactions