missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Transcend™ Nonradioactive Translation Detection Systems: IRNA
Description
- Non-Radioactive Detection of In Vitro Translated Proteins
- Biotin tag allows detection of 0.5-5ng of protein 3-4 hours after gel electrophoresis
- No radioisotope handling, storage or disposal issues
- Option of colorimetric or chemiluminescent detection
Specifications
Specifications
| Content And Storage | <-65°C |
| For Use With (Application) | Non-radioactive detection of Proteins synthesized in-vitro |
| Includes | Transcend tRNA |
| Product Type | Transcend tRNA |
| Quantity | 30 x 50 μL Reactions |