missing translation for 'onlineSavingsMsg'
Learn More

Promega ToxGlo™ Mitochondrial Toxicity Assay

The Mitochondrial ToxGlo™ Assay provides a method for predicting potential mitochondrial dysfunction as a result of xenobiotic exposure.

$478.29 - $2,603.57

Specifications

Format Bottle
Content And Storage -30°C to -10°C
Reaction Time <1 Hour
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRG8000
Encompass
View Documents Promotion Details
Promega
G8000
10 mL
Each for $478.29
Only null left
 
PRG8001
Encompass
View Documents Promotion Details
Promega
G8001
100 mL
Each for $2,603.57
Only null left
 
Description

Description

  • A Cell-Based Method for Predicting Potential Mitochondrial Dysfunction
  • Distinguish primary mitochondrial dysfunction from secondary cytotoxic events
  • Predictive for mitochondrial toxicities
  • Fast, cost-effective and easy to implement
Specifications

Specifications

Bottle
-30°C to -10°C
<1 Hour
Videos
SDS
Documents

Documents