missing translation for 'onlineSavingsMsg'
Learn More

Promega TNT™ T7 Quick for PCR DNA

Catalog No. PRL5540
Encompass
Change view
Click to view available options
Quantity:
40 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL5540 40 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRL5540 Supplier Promega Supplier No. L5540

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Quick Master Mix of reagents which contains recombinant RNasin™ ribonuclease inhibitor

TNT T7 Quick for PCR DNA is a rapid, convenient, coupled transcription/translation system designed for optimum protein expression from PCR templates.

  • Coupled Transcription/Translation System for Optimum Protein Expression from PCR Templates
  • Express protein directly from PCR, no cleanup needed
  • Up to 5 times more protein than standard translation reactions with linear templates
  • Includes Recombinant RNasin(R) Ribonuclease Inhibitor

  • U.S. Patents 5,324,637 and 5,492,817, Australian Patent 660329 and other patents.
  • U.S. Patent 5,552,302, Australian Patent 646803 and other patents.

Specifications

Content And Storage <-65°C
For Use With (Application) One-tube reaction for optimum expression of PCR templates
Includes 8 x 200μL TNT™ T7 PCR quick master mix, 1 x 50μL Methionine, 1 x 1.25mL Nuclease-free Water
No. of Reactions 40
Quantity 40 Reactions