missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega TNT™ Coupled Reticulocyte Lysate Systems
TNT Coupled Reticulocyte Lysate Systems are single-tube, coupled transcription/translation systems for eukaryotic cell-free protein expression.
$374.20 - $1404.56
Specifications
| Content And Storage | <-65°C |
|---|---|
| For Use With (Application) | GST pull-downs, Gel shift assays, co-immunoprecipitation, characterization of Protein modifications, Protein-RNA interactions, Protein activity studies, confirmation of gene products, Protein truncation test (PTT) |
Includes: TNT Rabbit Reticulocyte Lysate, Appropriate TNT polymerase, Control DNA, TNT Reaction Buffer, Luciferase assay wells, Luciferase assay reagent, Amino acid mixtures
| Catalog Number | Mfr. No. | Product Type | No. of Reactions | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | No. of Reactions | Price | Quantity | ||||
PR-L4600
|
Promega
L4600 |
TnT SP6 Coupled Reticulocyte Lysate System | 40 |
Each for $1,350.55
|
|
||||
PR-L4610
|
Promega
L4610 |
TnT T7 Coupled Reticulocyte Lysate System | 40 |
Each for $1,357.13
|
|
||||
PR-L5010
|
Promega
L5010 |
TnT T7/T3 Coupled Reticulocyte Lysate System | 40 |
Each for $1,357.13
|
|
||||
PR-L4950
|
Promega
L4950 |
TnT T3 Coupled Reticulocyte Lysate System | 40 |
Each for $1,357.13
|
|
||||
PR-L5020
|
Promega
L5020 |
TnT T7/SP6 Coupled Reticulocyte Lysate System | 40 |
Each for $1,404.56
|
|
||||
PR-L4611
|
Promega
L4611 |
TnT T7 Coupled Reticulocyte Lysate System | 8 Reactions - Trial Size |
Each for $374.20
|
|
||||
PR-L4601
|
Promega
L4601 |
TnT SP6 Coupled Reticulocyte Lysate System | 8 Reactions - Trial Size |
Each for $374.20
|
|
||||
Description
- A Single-Tube, Coupled Transcription/Translation System
- Study protein:protein, protein:nucleic acid and protein expression interactions
- Generate proteins in one hour versus days
- You choose the promoter
- U.S. Patents 5,324,637 and 5,492,817; Australian Patent 660329.
- U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289.
- Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
Specifications
| <-65°C | |
| GST pull-downs, Gel shift assays, co-immunoprecipitation, characterization of Protein modifications, Protein-RNA interactions, Protein activity studies, confirmation of gene products, Protein truncation test (PTT) |