Learn More
Promega T7 RiboMAX™ Express RNAi System
Catalog No. PRP1700
Encompass
Change view
Click to view available options
Quantity:
50 x 20 μL
Catalog No. Quantity
PRP1700 50 x 20 μL
1 options
Catalog No. PRP1700 Supplier Promega Supplier No. P1700
Only null left
Includes
T7 Express Enzyme Mix; RiboMAX Express T7 2X Buffer; pGEM Express Positive Control Template; Sodium Acetate, 3.0M (pH 5.2); RNase A Solution; RQ1 RNase-free DNase; Nuclease-free Water
The T7 RiboMAX Express RNAi System is an in vitro transcription system designed to produce milligram amounts of RNA suitable for use in RNAi assays.
  • Produces Milligam Amounts of RNA for RNA Interference Experiments
  • siRNA duplexes for mammalian RNAi
  • Long dsRNA for nonmammalian RNAi
  • Produces milligram amounts of double-stranded RNA in 30 minutes
  • U.S. Patent 5,552,302.
  • This product is covered under license from Carnegie Institution of Washington under U.S. Patent 6,506,559, Australian Patent 743798 and other patents pending. Commercial use of this product will require a separate license from Carnegie.
Specifications
Content And Storage -30°C to -10°C
For Use With (Application) Synthesis of siRNA duplexes for mammalian RNA interference, generation of large quantities of long dsRNA for non-mammalian RNA interference
Includes 100μL Enzyme mix, T7 Express (T7 RNA Polymerase, recombinant RNasin™ RNase inhibitor and recombinant inorganic Pyrophosphatase), 500μL RiboMAX™ Express T7 2X Buffer, 110U 1U/
Quantity 50 x 20 μL