missing translation for 'onlineSavingsMsg'
Learn More

Promega rNTPs

Catalog No. PRE6021
Encompass
Change view
Click to view available options
Product Type:
rATP
rCTP
rGTP
rNTP Set (rCTP, rATP, rUTP, rGTP)
rUTP
Quantity:
400 μL
4 x 400 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PRE6021 rUTP 400 μL
PRE6000 rNTP Set (rCTP, rATP, rUTP, rGTP) 4 x 400 μL
PRE6011 rATP 400 μL
PRE6041 rCTP 400 μL
PRE6031 rGTP 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PRE6021 Supplier Promega Supplier No. E6021
Only null left

High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.

  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400μL rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC

Specifications

Product Type rUTP
Content And Storage -30°C to -10°C
Concentration 100 mM
Quantity 400 μL
For Use With (Application) Provides a cell-free system for in-vitro transcription of eukaryotic genes