Learn More
Promega RNA Markers
RNA Markers consist of a ladder of 9 RNA transcripts synthesized in vitro from specific templates. Sizes: 281, 623, 955, 1,383, 1,908, 2,604, 3,638, 4,981 and 6,583 bases.
Supplier: Promega G3191
Description
- Ladder of Nine RNA Transcripts 281-6,583 Bases
- Recommended loading: 3μL (prepared in formaldehyde/MOPS buffer and separated onto a 1% formaldehyde-agarose gel using MOPS running buffer)
Specifications
| 1 μg/μL | |
| Liquid | |
| Size estimation of single stranded RNA from 0.28 to 6.58kb in glyoxal or Formaldehyde Agarose gels | |
| 50 μL |
| RNA Molecular Weight Marker/Ladder | |
| Prepared in Formaldehyde/MOPS Buffer and separated onto a 1% Formaldehyde-Agarose Gel using MOPS running Buffer | |
| 9 | |
| <−65°C |