Learn More
Promega RiboMAX™ Large Scale RNA Production Systems
Catalog No. PRP1280
Encompass
Change view
Click to view available options
Product Type:
RiboMAX Large Scale RNA Production System-SP6
Catalog No. Product Type
PRP1280 RiboMAX Large Scale RNA Production System-SP6
1 options
Catalog No. PRP1280 Supplier Promega Supplier No. P1280
Only null left
Includes
Linear control DNA; RQ1 RNase-free DNase; Nuclease-free water; Enzyme mix; rATP; rCTP; rGTP; rUTP; Sodium acetate, 3.0M (pH 5.2); Transcription buffer, 5X
Designed for large-scale RNA production, the RiboMAX Systems consistently produce 2-5mg RNA from a 1ml reaction volume.
  • Capped or Uncapped RNA Transcripts for in vitro or in vivo Translation
  • Produces 2-5mg/mL RNA in a 1mL reaction
  • Available for use with SP6 or T7 RNA polymerase
  • Scalable to meet varying RNA production requirements
  • U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
  • The RiboMAX Large Scale RNA Production System—T7 (Mfr. No. P1300) is covered by U.S. Patents 5,256,555; 6,586,218; 6,586,219 and other pending patents and is sold under a license from Ambion, Inc. It is intended for research use only. Parties wishing to use this product for other applications should contact Ambion, Inc.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
Content And Storage -30°C to -10°C
For Use With (Application) Produce 2 to 5mg/mL of RNA in a 1mL reaction, about 10- to 20-fold more RNA than is produced with the standard Riboprobe™ system transcription reaction
Includes Linear control DNA, RQ1 RNase-free DNase, Nuclease-free Water, Enzyme mix, rATP, rCTP, rGTP, rUTP, Sodium Acetate, 3m (pH 5.2), 5X Transcription Buffer
Product Type RiboMAX Large Scale RNA Production System-SP6
Quantity 1 System