Learn More
Promega Ribo m7G Cap Analog DFS Item
This methylated ribonucleotide can be incorporated onto the 5'-end of in vitro transcripts and simulates the cap structure found on most eukaryotic mRNA molecules.
Supplier:  Promega P1712
 View more versions of this product
Catalog No. PR-P1712
Only null left
Explore available promotions
Description
Description
  • Enhances Translation Efficiency in Reticulocyte-Based Reactions
  • Protects RNA from intracellular digestion
  • Can be used with Riboprobe(R) or RiboMAX™ RNA production systems
  • Supplied at a concentration of 40mM
Specifications
Specifications
-30°C to -10°C
Contains at least 34.3μL Ribo m7G cap analog in Nuclease-free Water
Enhances translation efficiency in many reticulocyte-based reactions and protects RNA from intracellular digestion
25 A254 U
Product Suggestions
Product Suggestions