missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Rabbit Reticulocyte Lysate, Untreated
Description
- Characterize Protein Products, Study Transcriptional and Translational Control
- Contains tRNA, ribosomes, amino acids, initiation, elongation and termination factors needed for protein synthesis
- No micrococcal nuclease treatment
- Consistent reticulocyte production from every lot
Specifications
Specifications
| Content And Storage | <-65°C |
| For Use With (Application) | Isolation of the cellular components necessary for Protein synthesis |
| Quantity | 1 mL |