missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Rabbit Reticulocyte Lysate Systems, Nuclease Treated
Description
Includes
Luciferase control RNA; Reticulocyte lysate, nuclease treated; Amino acid mixtures
- In Vitro Translation of mRNA and Characterization of mRNA Products
- Optimized for translation and ready to use
- Contains an energy-regenerating system, a mix of tRNAs, hemin, potassium chloride and magnesium acetate
- U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
- Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
Specifications
| Content And Storage | <-65°C |
| For Use With (Application) | Identification of mRNA species, the characterization of their Protein products and the investigation of transcriptional and translational control |
| Includes | 1mL Rabbit reticulocyte lysate, Nuclease treated (5 x 200μL), 1mM 50μL Amino Acid mixture minus Methionine, 1mM 50μL Amino Acid mixture minus Leucine, 1mM 50μL Amino Acid mixture minu |
| Quantity | 5 x 200 μL |