missing translation for 'onlineSavingsMsg'
Learn More

Promega pGEM™ Conventional DNA Digest Marker

Catalog No. PRG1741
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG1741 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG1741 Supplier Promega Supplier No. G1741

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

6X Blue/Orange Loading Dye

Fifteen phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 36bp to 2,645bp. These markers are generated from separate digests of pGEM(R)-3 Vector DNA with HinfI, RsaI and AvaII and then combined.

  • Fifteen DNA Fragments of 36-2,645bp, Combined From Three Separate Digests of pGEM(R)-3 Vector
  • Load 1μL/lane
  • Supplied at a concentration of 1μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.8, 1mM EDTA
Color Blue, Orange
Concentration 1 μg/μL
Form Liquid
Formulation 50μg pGEM™ DNA marker, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 15
Quantity 50 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker