missing translation for 'onlineSavingsMsg'
Learn More

Promega P450-Glo™ CYP450 Assay Systems

Catalog No. PRV8792
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
Product Type:
P450-Glo CYP1A1 Assay
P450-Glo CYP1A2 Assay
P450-Glo CYP1B1 Assay
P450-Glo CYP2C19 Screening Assay
P450-Glo CYP2C8 Assay
P450-Glo CYP2C9 Assay
P450-Glo CYP2D6 Screening Assay
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV8792 50 mL P450-Glo CYP2C9 Assay
PRV8772 50 mL P450-Glo CYP1A2 Assay
PRV8882 50 mL P450-Glo CYP2C19 Screening Assay
PRV8892 50 mL P450-Glo CYP2D6 Screening Assay
PRV8751 10 mL P450-Glo CYP1A1 Assay
PRV8752 50 mL P450-Glo CYP1A1 Assay
PRV8761 10 mL P450-Glo CYP1B1 Assay
PRV8771 10 mL P450-Glo CYP1A2 Assay
PRV8791 10 mL P450-Glo CYP2C9 Assay
PRV8762 50 mL P450-Glo CYP1B1 Assay
PRV8891 10 mL P450-Glo CYP2D6 Screening Assay
PRV8781 10 mL P450-Glo CYP2C8 Assay
PRV8881 10 mL P450-Glo CYP2C19 Screening Assay
PRV8782 50 mL P450-Glo CYP2C8 Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. PRV8792 Supplier Promega Supplier No. V8792

Includes

Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA

Provide a complete set of reagents for performing luminescent cytochrome P450 assays

  • A Luminescent Method and Screening System for Measuring Activity
  • Screening system includes complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background

Specifications

Quantity 50 mL
Content And Storage 1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer, 1 x 1mL Luciferin-H 5mM -30°C to -10°C
Detection Method Homogeneous, Luminescence
For Use With (Application) For measuring cytochrome P450 activity
Product Type P450-Glo CYP2C9 Assay

  • U.S. Patent 6,602,677; Australian Pat. No. 754312 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673.