missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega P450-Glo™ CYP1A2 Induction/Inhibition Assay
The P450-Glo™ CYP1A2 Assay and Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays
Supplier: Promega V8421
Includes: Luciferin-1A2 (30μL), 500X D-Cysteine (100μL), luciferin detection reagent and reconstitution buffer.
Description
- A Luminescent Assay and Screening System for Measuring CYP1A2 Activity
- Screening system includes a complete set of reagents
- Luminescent format eliminates need for time-consuming analyses
- Broad dynamic range and low background
Specifications
| 10 mL | |
| Homogeneous, Luminescence | |
| 1 x 30μL Luciferin-1A2, 1 x 100μL D-Cysteine 500X, 1 x 1 each Luciferin detection reagent, 1 x 10mL Reconstitution Buffer |
| -30°C to -10°C | |
| For measuring cytochrome P450 activity | |
| For 384-well format |