Learn More
Promega NADP/NADPH-Glo™ Assay
The NAD/NADH-Glo™ Assay is a bioluminescent, homogeneous single-reagent-addition assay for detecting total oxidized and reduced nicotinamide adenine dinucleotides and determining their ratio in biological samples or in defined enzyme reactions.
Supplier: Promega G9082
Includes:
- G9081 assay kit components: Reductase (55μL), Reductase Substrate (55μL), NADP Cycling Enzyme (1 vial), NADP Cycling Substrate (275μL), Luciferin Detection Reagent; Reconstitution Buffer (10mL)
- G9082 assay kit components:Reductase (275μL), Reductase Substrate (275μL), NADP Cycling Enzyme (1 vial), NADP Cycling Substrate (275μL), Luciferin Detection Reagent; Reconstitution Buffer (50mL)
Description
- Bioluminescent Assays to Detect NAD(P)+ and NAD(P)H Levels in Cells
- Homogeneous, single-reagent-addition assays
- Easily adapted for high throughput screening formats
Chemical Identifiers
| NADP/NADPH-Glo assay kit |
Specifications
| 50 mL | |
| NADP/NADPH-Glo assay kit |