missing translation for 'onlineSavingsMsg'
Learn More

Promega NADP/NADPH-Glo™ Assay

Catalog No. PRG9081
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9081 10 mL
PRG9082 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG9081 Supplier Promega Supplier No. G9081
Only null left

Includes

  • G9081 assay kit components: Reductase (55μL), Reductase Substrate (55μL), NADP Cycling Enzyme (1 vial), NADP Cycling Substrate (275μL), Luciferin Detection Reagent; Reconstitution Buffer (10mL)
  • G9082 assay kit components:Reductase (275μL), Reductase Substrate (275μL), NADP Cycling Enzyme (1 vial), NADP Cycling Substrate (275μL), Luciferin Detection Reagent; Reconstitution Buffer (50mL)

The NAD/NADH-Glo™ Assay is a bioluminescent, homogeneous single-reagent-addition assay for detecting total oxidized and reduced nicotinamide adenine dinucleotides and determining their ratio in biological samples or in defined enzyme reactions.

  • Bioluminescent Assays to Detect NAD(P)+ and NAD(P)H Levels in Cells
  • Homogeneous, single-reagent-addition assays
  • Easily adapted for high throughput screening formats

Specifications

Quantity 10 mL
Product Type NADP/NADPH-Glo assay kit