missing translation for 'onlineSavingsMsg'
Learn More

Promega MultiTox-Glo™ Multiplex Cytotoxicity Assay

Catalog No. PRG9272
Encompass
Change view
Click to view available options
Quantity:
10 mL
2 x 50 mL
5 x 10 mL
No. of Reactions:
100 x 96-well, 400 x 384-well
1000 x 96-well and 4000 x 384-well
500 x 96-well and 2000 x 384-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity No. of Reactions
PRG9272 2 x 50 mL 1000 x 96-well and 4000 x 384-well
PRG9271 5 x 10 mL 500 x 96-well and 2000 x 384-well
PRG9270 10 mL 100 x 96-well, 400 x 384-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG9272 Supplier Promega Supplier No. G9272

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Assay buffer, AAF-Glo™ substrate, GF-AFC substrate

The MultiTox-Glo™ Multiplex Cytotoxicity Assay is a fluorescent and luminescent assay that measures the relative number of live and dead cells in a population.

  • Measures Relative Number of Live and Dead Cells
  • Fluorescent and luminescent assay
  • Normalize data with a built-in internal control
  • Immediately identify more false-positives and false-negatives

Specifications

Quantity 2 x 50 mL
Content And Storage -30°C to -10°C
No. of Reactions 1000 x 96-well and 4000 x 384-well
Detection Method Fluorescence, Luminescence

  • Patent Pending.
  • U.S. Patents 6,602,677 and 7,241,584, Australian Patents 754312 and 785294 and other patents and patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.