missing translation for 'onlineSavingsMsg'
Learn More

Promega Molecular Grade Ethidium Bromide Solution

Catalog No. PRH5041
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRH5041 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRH5041 Supplier Promega Supplier No. H5041

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A fluorescent dye suitable for staining nucleic acids after electrophoresis or in cesium chloride gradients.

  • Quality tested and certified free of DNase, RNase and protease activity
  • Supplied at a concentration of 10mg/ml

Specifications

Color Dark Red/Purple
Concentration 10 mg/mL
Form Liquid
For Use With (Application) Staining Nucleic Acids after electrophoresis
Quantity 10 mL
Storage Requirements 15 to 30°C
Product Type Fluorescent DNA/RNA Stain
Stain Type Fluorescent