missing translation for 'onlineSavingsMsg'
Learn More

Promega M-MLV Reverse Transcriptase

Catalog No. PRM1701
Encompass
Change view
Click to view available options
pH:
7.5
8.3
Description:
M-MLV Reverse Transcriptase
Quantity:
10000U
50000U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Description Quantity
PRM1701 8.3 M-MLV Reverse Transcriptase 10000U
PRM1705 7.5 M-MLV Reverse Transcriptase 50000U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRM1701 Supplier Promega Supplier No. M1701
Only null left

M-MLV RT is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb).

  • RNA-Dependent DNA Polymerase for cDNA Synthesis
  • Enzyme supplied at a concentration of 200u/μL

Specifications

Concentration 200 U/μL
Content And Storage −30 to −10°C
Description M-MLV Reverse Transcriptase
Form Liquid
pH 8.3
Quantity 10000U
For Use With (Application) cDNA Synthesis
Includes 5X reaction buffer (250mM Tris-HCl [pH 8.3 at 25°C], 375mM KCl, 15mM MgCl2, 50mM DTT), buffer also available separately
Source Recombinant E. coli Strain
Storage Buffer Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 0.2M, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50%