missing translation for 'onlineSavingsMsg'
Learn More

Promega φX174 DNA/HaeIII Conventional DNA Digest Marker

Catalog No. PRG1761
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG1761 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG1761 Supplier Promega Supplier No. G1761
Only null left

Includes

6X Blue/Orange Loading Dye

Eleven phenol-extracted, ethanol-precipitated DNA fragments ranging in size from 72bp to 1,353bp.

  • Eleven DNA Fragments of 72-1,353bp
  • Load 1μL/lane
  • Supplied at a concentration of 1μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.5, 1mM EDTA
Color Blue, Orange
Concentration 1 μg/μL
Form Liquid
Formulation 50μg X174 DNA/HaeIII marker, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 11
Quantity 50 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker