missing translation for 'onlineSavingsMsg'
Learn More

Promega Lambda DNA

Catalog No. PRD1501
Encompass
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD1501 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRD1501 Supplier Promega Supplier No. D1501

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Generates molecular weight size markers used in gel analysis of nucleic acids; nucleotide sequence has been determined.

  • Also commonly used as a substrate in restriction enzyme activity assays

Specifications

Concentration 0.55 μg/μL
Form Liquid
For Use With (Application) For used as a substrate in restriction Enzyme activity assays
No. of Bands 1
Quantity 250 μg
Storage Requirements −30 to −10°C
Product Type Lambda DNA