missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Kinase-Glo™ Max Luminescent Kinase Assay
Description
- Monitors Activity of Purified Kinases by Quantifying ATP
- Luminescent signal is inversely proportional to the amount of kinase activity
- Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP
- U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024, 5,674,713 and 5,700,673.
Specifications
Specifications
| Quantity | 10 mL |
| Content And Storage | 1 x 10mL Kinase-Glo™ max Buffer, 1 x resuspend to 10mL Kinase-Glo™ max substrate. -30°C to -10°C |
| Detection Method | Homogeneous, Non-radioactive Method |
| For Use With (Application) | For determining the activity of purified Kinases by quantifying the amount of ATP |
| Product Type | Kinase-Glo Max Luminescent Kinase Assay |
| For Use With (Equipment) | For multiwell plates |