missing translation for 'onlineSavingsMsg'
Learn More

Promega Kinase-Glo™ Luminescent Kinase Assay

The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.

Supplier:  Promega V6714

Encompass

 View more versions of this product

Catalog No. PRV6714


Only null left

Description

Description

  • Monitors Activity of Purified Kinases by Quantifying ATP
  • Luminescent signal is inversely proportional to the amount of kinase activity
  • Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP
Specifications

Chemical Identifiers

-20°C
For determining the activity of purified Kinases by quantifying the amount of ATP
For multiwell plates
Homogeneous, Non-radioactive Method
Kinase-Glo Luminescent Kinase Assay

Specifications

10 x 100 mL
Homogeneous, Non-radioactive Method
Kinase-Glo Luminescent Kinase Assay
-20°C
For determining the activity of purified Kinases by quantifying the amount of ATP
For multiwell plates
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions