missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Kinase-Glo™ Luminescent Kinase Assay
The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.
Supplier: Promega V6712
Description
- Monitors Activity of Purified Kinases by Quantifying ATP
- Luminescent signal is inversely proportional to the amount of kinase activity
- Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP
Chemical Identifiers
| -20°C | |
| For determining the activity of purified Kinases by quantifying the amount of ATP | |
| For multiwell plates |
| Homogeneous, Non-radioactive Method | |
| Kinase-Glo Luminescent Kinase Assay |
Specifications
| 10 x 10 mL | |
| Homogeneous, Non-radioactive Method | |
| Kinase-Glo Luminescent Kinase Assay |
| -20°C | |
| For determining the activity of purified Kinases by quantifying the amount of ATP | |
| For multiwell plates |