missing translation for 'onlineSavingsMsg'
Learn More

Promega Kemptide (PKA) Peptide Substrate

Catalog No. PRV5601
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV5601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV5601 Supplier Promega Supplier No. V5601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kemptide is a peptide substrate for cAMP-Dependent Protein Kinase with the sequence Leu-Arg-Arg-Ala-Ser-Leu-Gly.

  • Specific Substrate for cAMP-Dependent Protein Kinases
  • Supplied ready for use in kinase reactions

Specifications

Purity ≥70%
Concentration 10 mg/mL
Content And Storage -30°C to -10°C
Molecular Weight (g/mol) 772
Product Type Kemptide (PKA) Peptide Substrate
Quantity 1 mg
Sequence Leu-Arg-Arg-Ala-Ser-Leu-Gly