missing translation for 'onlineSavingsMsg'
Learn More

Promega E. coli T7 S30 Extract System for Circular DNA

Catalog No. PRL1130
Encompass
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRL1130 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRL1130 Supplier Promega Supplier No. L1130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

PinPoint™ Xa Control Vector, 5μg; T7/S30 extract for circular DNA, 3 x 150μL; S30 premix without amino acids, 750μL; Amino acid mixture minus methionine, 175μL; Amino acid mixture minus cysteine, 175μL; Amino acid mixture minus leucine, 175μL

The E. coli T7 S30 Extract System for Circular DNA simplifies the transcription/translation of DNA sequences cloned in plasmid or lambda vectors containing a T7 promoter.

  • Create Radiolabeled Protein for Transcription/Translation Studies
  • Translate using clones with a T7 promoter and a ribosome binding site; no E. coli promoter required
  • Reduce the chance of expressed proteins degrading
  • Includes all components needed for coupled transcription/translation

For research purposes only. Not for diagnostic or therapeutic use. For nonresearch uses of the portion of the vector encoding the biotinylation sequence, please contact Promega Corporation for licensing information.

Specifications

Content And Storage <-65°C
For Use With (Application) Synthesis of radiolabeled Protein, verification of cloned gene expression before in-vivo Protein expression in E. coli, use of Protein in functional studies of transcription and translation, use of Protein as a tracer in Protein purification, incorporation of un-natural Amino Acids into Proteins for structural studies
Includes 5μg PinPoint Xa control vector, 3 x 150μL T7/S30 extract for circular DNA, 750μL S30 premix without Amino Acids, 175μL Amino Acid mixture minus Methionine, 175μL Ami
No. of Reactions 30
Quantity 30 Reactions