missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GSH/GSSG-Glo™ Glutathione Assay
Description
- A Luminescent Assay for the Detection and Quantification of Total Glutathione Ratios
- Measures physiologically relevant GSH/GSSG ratios
- Simple protocol minimizes sample handling, reducing variability
Specifications
Specifications
| Quantity | 50 mL |
| Content And Storage | 1 x 1.25mL DTT 100mM, 1 x 500μL Luciferin-NT, 3 x 500μL Glutathione S-transferase solution, 1 x 500μL Glutathione 5mM, 1 x 1 each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer with Estera -30°C to -10°C |
| For Use With (Application) | For the detection and quantification of total Glutathione (GSH +GSSG), GSSG and GSH/GSSG ratios in cultured cells |
| For Use With (Equipment) | For rapid multiwell plate |