missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GSH-Glo™ Glutathione Assay
Description
Includes
Reconstitution Buffer with esterase, Luciferin-NT, GSH-Glo™ Reaction Buffer, Glutathione S-Transferase, Glutathione (5mM), Luciferin Detection Reagent
- A Luminescent-Based Assay for the Detection and Quantification of Glutathione (GSH)
- Measure reduced and/or total glutathione levels in cells and tissues
- Measure GSH levels directly from cell culture wells
- Excellent sensitivity and easily scalable
Specifications
Specifications
| Quantity | 50 mL |
| Content And Storage | 1 x 50mL Reconstitution Buffer with Esterase, 1 x 50μL Luciferin-NT, 1 x 50mL GSH-Glo Reaction Buffer, 1 x 500μL Glutathione S-transferase, 1 x 100μL Glutathione 5mM, 1 x 1 each Luci. -30°C to -10°C |
| For Use With (Application) | For detection and quantification of Glutathione in cells or in various biological samples |