missing translation for 'onlineSavingsMsg'
Learn More

Promega GSH-Glo™ Glutathione Assay

Catalog No. PRV6911
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6911 10 mL
PRV6912 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV6911 Supplier Promega Supplier No. V6911
Only null left

Includes

Reconstitution Buffer with esterase, Luciferin-NT, GSH-Glo™ Reaction Buffer, Glutathione S-Transferase, Glutathione (5mM), Luciferin Detection Reagent

The GSH-Glo™ Assay is a luminescent-based assay for the detection and quantification of glutathione (GSH) in cells or in various biological samples.

  • A Luminescent-Based Assay for the Detection and Quantification of Glutathione (GSH)
  • Measure reduced and/or total glutathione levels in cells and tissues
  • Measure GSH levels directly from cell culture wells
  • Excellent sensitivity and easily scalable

Specifications

Quantity 10 mL
Content And Storage -30°C to -10°C
For Use With (Application) For detection and quantification of Glutathione in cells or in various biological samples
Kit Contents 1 x 10mL Reconstitution Buffer with Esterase, 1 x 100μL Luciferin-NT, 1 x 10mL GSH-Glo Reaction Buffer, 1 x 100μL Glutathione S-transferase, 1 x 100μL Glutathione 5mM, 1 x 1 each Luc